3-Fluor-N-methylanilin
Lieferant: ACTIVATE SCIENTIFIC
AS35439G1
AS35439G5
ACTIAS35439G1EA
0
CHF
ACTIAS35439G1
ACTIAS35439G5
3-Fluor-N-methylanilin
3-Fluor-N-methylanilin
Formel:
C₇H₈FN MW: 125,15 g/mol |
MDL Number:
MFCD02683095 CAS-Nummer: 1978-37-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...