5-Fluor-m-toluonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32313G1 AS32313G10 AS32313G5
ACTIAS32313G1EA 0 CHF
ACTIAS32313G1 ACTIAS32313G10 ACTIAS32313G5
5-Fluor-m-toluonitril
5-Fluor-m-toluonitril
Formel: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD03094312
CAS-Nummer: 216976-30-6
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR