3-Fluor-o-toluonitril
Lieferant: ACTIVATE SCIENTIFIC
AS34970G1
AS34970G10
AS34970G5
ACTIAS34970G1EA
0
CHF
ACTIAS34970G1
ACTIAS34970G10
ACTIAS34970G5
3-Fluor-o-toluonitril
3-Fluor-o-toluonitril
Formel:
C₈H₆FN MW: 135,14 g/mol |
MDL Number:
MFCD03788541 CAS-Nummer: 185147-06-2 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...