2-Fluor-p-toluonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32490G1 AS32490G5
ACTIAS32490G1EA 0 CHF
ACTIAS32490G1 ACTIAS32490G5
2-Fluor-p-toluonitril
2-Fluor-p-toluonitril
Formel: C₈H₆FN
MW: 135,14 g/mol
MDL Number: MFCD03094311
CAS-Nummer: 85070-67-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR