3-Chlorbenzoylacetonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3'-Chloro-2-cyanoacetophenone, (3-Chlorobenzoyl)acetonitrile, 3-(3-Chlorophenyl)-3-oxopropanenitrile

AS34862G1 AS34862G10 AS34862G5
ACTIAS34862G1EA 0 CHF
ACTIAS34862G1 ACTIAS34862G10 ACTIAS34862G5
3-Chlorbenzoylacetonitril
3-Chlorbenzoylacetonitril
Formel: C₉H₆ClNO
MDL Number: MFCD00067891
CAS-Nummer: 21667-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR