2-(2-Chlorphenyl)ethanol

Lieferant: ACTIVATE SCIENTIFIC

AS21644G25 AS21644G5
ACTIAS21644G25EA 0 CHF
ACTIAS21644G25 ACTIAS21644G5
2-(2-Chlorphenyl)ethanol
2-(2-Chlorphenyl)ethanol
Formel: C₈H₉ClO
MW: 156.61 g/mol
MDL Number: MFCD00002888
CAS-Nummer: 19819-95-5
EINECS: 243-348-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR