3-Brom-o-toluonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32616G1 AS32616G5
ACTIAS32616G1EA 0 CHF
ACTIAS32616G1 ACTIAS32616G5
3-Brom-o-toluonitril
3-Brom-o-toluonitril
Formel: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD09025665
CAS-Nummer: 52780-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR