1-Brom-5-chlor-2-iodbenzol

Lieferant: ACTIVATE SCIENTIFIC

AS32794G1 AS32794G5
ACTIAS32794G1EA 0 CHF
ACTIAS32794G1 ACTIAS32794G5
1-Brom-5-chlor-2-iodbenzol
1-Brom-5-chlor-2-iodbenzol
Formel: C₆H₃BrClI
MW: 317,35 g/mol
MDL Number: MFCD00672944
CAS-Nummer: 31928-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR