2-Brombenzoylacetonitril

Lieferant: ACTIVATE SCIENTIFIC

AS25008G25 AS25008G5
ACTIAS25008G25EA 0 CHF
ACTIAS25008G25 ACTIAS25008G5
2-Brombenzoylacetonitril
2-Brombenzoylacetonitril
Formel: C₉H₆BrNO
MW: 224.06 g/mol
CAS-Nummer: 53882-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR