4-Chlor-m-anisidin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Amino-2-chloroanisole, 4-Chloro-3-methoxyaniline, 4-Chloro-3-anisidine
AS22644G1
AS22644G25
AS22644G5
ACTIAS22644G1EA
0
CHF
ACTIAS22644G1
ACTIAS22644G25
ACTIAS22644G5
4-Chlor-m-anisidin
4-Chlor-m-anisidin
Formel:
C₇H₈ClNO MW: 157.6 g/mol |
MDL Number:
MFCD00672967 CAS-Nummer: 13726-14-2 UN: 2233 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...