3-Methoxy-p-toluidin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
1-Amino-3-methoxy-4-methylbenzene, 4-Amino-2-methoxytoluene, 3-Methoxy-4-methylaniline, 4-Methyl-m-anisidine, 5-Amino-2-methylanisole
AS33718G1
AS33718G5
ACTIAS33718G1EA
0
CHF
ACTIAS33718G1
ACTIAS33718G5
3-Methoxy-p-toluidin
3-Methoxy-p-toluidin
Formel:
C₈H₁₁NO MW: 137.18 g/mol Siedepunkt: 250 Melting Pt: 58 |
MDL Number:
MFCD00025371 CAS-Nummer: 16452-01-0 EINECS: 240-500-8 UN: 2811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...