3-Fluor-o-anisidin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Fluoro-2-methoxyaniline, 3-Fluoro-2-anisidine, 2-Amino-6-fluoroanisole
AS33601G1
AS33601G10
AS33601G5
ACTIAS33601G1EA
0
CHF
ACTIAS33601G1
ACTIAS33601G10
ACTIAS33601G5
3-Fluor-o-anisidin
3-Fluor-o-anisidin
Formel:
C₇H₈FNO MW: 141.15 g/mol |
MDL Number:
MFCD00007759 CAS-Nummer: 437-83-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...