4-(Trifluormethoxy)-o-toluidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Amino-5-(trifluoromethoxy)toluene, 4-Trifluoromethoxy-o-toluidine, 4-(Trifluoromethoxy)-2-toluidine

AS35213G1 AS35213G5
ACTIAS35213G1EA 0 CHF
ACTIAS35213G1 ACTIAS35213G5
4-(Trifluormethoxy)-o-toluidin
4-(Trifluormethoxy)-o-toluidin
Formel: C₈H₈F₃NO
MW: 191,15 g/mol
MDL Number: MFCD01631541
CAS-Nummer: 86256-59-9
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR