6-(Trifluormethyl)-o-anisidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Amino-3-methoxybenzotrifluoride, 2-Amino-3-(trifluoromethyl)anisole, 6-(Trifluoromethyl)-2-anisidine, 6-(Trifluoromethyl)-2-methoxyaniline, 2-Methoxy-6-(trifluoromethyl)aniline

AS34563G1 AS34563G5
ACTIAS34563G1EA 0 CHF
ACTIAS34563G1 ACTIAS34563G5
6-(Trifluormethyl)-o-anisidin
6-(Trifluormethyl)-o-anisidin
Formel: C₈H₈F₃NO
MW: 191.15 g/mol
MDL Number: MFCD09832384
CAS-Nummer: 133863-87-3
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR