6-Brom-o-toluidin

Lieferant: ACTIVATE SCIENTIFIC

AS34077G1 AS34077G5
ACTIAS34077G1EA 0 CHF
ACTIAS34077G1 ACTIAS34077G5
6-Brom-o-toluidin
6-Brom-o-toluidin
Formel: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD04039885
CAS-Nummer: 53848-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR