2-Brom-m-toluidin

Lieferant: ACTIVATE SCIENTIFIC

AS35493G1 AS35493G5
ACTIAS35493G1EA 0 CHF
ACTIAS35493G1 ACTIAS35493G5
2-Brom-m-toluidin
2-Brom-m-toluidin
Formel: C₇H₈BrN
MDL Number: MFCD06411367
CAS-Nummer: 54879-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR