2-(2,4-Dichlorphenyl)ethanol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,4-Dichlorophenethyl alcohol

AS35011G1 AS35011G10 AS35011G5
ACTIAS35011G1EA 0 CHF
ACTIAS35011G1 ACTIAS35011G10 ACTIAS35011G5
2-(2,4-Dichlorphenyl)ethanol
2-(2,4-Dichlorphenyl)ethanol
Formel: C₈H₈Cl₂O
MW: 191,06 g/mol
MDL Number: MFCD00192010
CAS-Nummer: 81156-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR