3,5-Dimethylbenzylbromid

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: α-Bromomesitylene, 1-(Bromomethyl)-3,5-dimethylbenzene

AS34631G10 AS34631G5
ACTIAS34631G10EA 0 CHF
ACTIAS34631G10 ACTIAS34631G5
3,5-Dimethylbenzylbromid
3,5-Dimethylbenzylbromid
Formel: C₉H₁₁Br
MW: 199,09 g/mol
Siedepunkt: 232 °C (1013 hPa)
Melting Pt: 40…41 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013539
CAS-Nummer: 27129-86-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR