1-Naphthylmethylamin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Naphthalenemethylamine, Naphthalen-1-ylmethanamine, Naphthalene-1-methylamine, 1-(Aminomethyl)naphthalene

AS35453G1 AS35453G5
ACTIAS35453G1EA 0 CHF
ACTIAS35453G1 ACTIAS35453G5
1-Naphthylmethylamin
1-Naphthylmethylamin
Formel: C₁₁H₁₁N
MW: 157,22 g/mol
Siedepunkt: 92 °C (0,1 mmHg)
Melting Pt: 267…269 °C
Dichte: 1,0958 g/cm³ (20 °C)
MDL Number: MFCD00004048
CAS-Nummer: 118-31-0
EINECS: 204-244-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR