1-(2-Methylphenyl)piperazin

Lieferant: ACTIVATE SCIENTIFIC

AS23862G100 AS23862G25
ACTIAS23862G100EA 0 CHF
ACTIAS23862G100 ACTIAS23862G25
1-(2-Methylphenyl)piperazin
1-(2-Methylphenyl)piperazin
Formel: C₁₁H₁₆N₂
MDL Number: MFCD00040729
CAS-Nummer: 39512-51-1
EINECS: 254-481-9
UN: 3263
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR