5-Methoxy-3-indolylessigsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Methoxy-3-indoleacetic acid, 2-(5-Methoxy-1H-indol-3-yl)acetic acid, 5-Methoxyindole-3-acetic acid
AS32350G1
AS32350G5
ACTIAS32350G1EA
0
CHF
ACTIAS32350G1
ACTIAS32350G5
5-Methoxy-3-indolylessigsäure
5-Methoxy-3-indolylessigsäure
Formel:
C₁₁H₁₁NO₃ MW: 205.21 g/mol Melting Pt: 150…151 °C |
MDL Number:
MFCD00005638 CAS-Nummer: 3471-31-6 EINECS: 222-438-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...