5-Brom-3-indolylessigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromoindole-3-acetic acid, 2-(5-Bromo-1H-indol-3-yl)acetic acid

AS31953G1 AS31953G5
ACTIAS31953G1EA 0 CHF
ACTIAS31953G1 ACTIAS31953G5
5-Brom-3-indolylessigsäure
5-Brom-3-indolylessigsäure
Formel: C₁₀H₈BrNO₂
Melting Pt: 174.5 °C
Dichte: 1.754 g/cm³ (20 °C)
MDL Number: MFCD00005637
CAS-Nummer: 40432-84-6
EINECS: 254-917-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR