(S)-(+)-Methylmandelat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: L(+)-Mandelic acid methyl ester, L(+)-2-Hydroxy-2-phenylacetic acid methyl ester, Methyl (S)-(+)-mandelate, L(+)-Methyl mandelate, (S)-(+)-Mandelic acid methyl ester, (S)-(+)-2-Hydroxy-2-phenylacetic acid methyl ester, L(+)-Methyl glycolate, (S)-(+)-Methyl glycolate

AS20992G100 AS20992G25
ACTIAS20992G100EA 0 CHF
ACTIAS20992G100 ACTIAS20992G25
(S)-(+)-Methylmandelat
(S)-(+)-Methylmandelat
Formel: C₉H₁₀O₃
Siedepunkt: 100…105 °C (1.25 mmHg)
Melting Pt: 58 °C
MDL Number: MFCD00064246
CAS-Nummer: 21210-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR