(R)-(-)-Methylmandelat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: D(-)-Methyl glycolate, (R)-(-)-Methyl glycolate, (R)-(-)-Mandelic acid methyl ester, D(-)-Methyl mandelate, D(-)-2-Hydroxy-2-phenylacetic acid methyl ester, D(-)-Mandelic acid methyl ester, (R)-Methyl 2-hydroxy-2-phenylacetate, Methyl (R)-(-)-mandelate, (R)-(-)-2-Hydroxy-2-phenylacetic acid methyl ester

AS20997G100 AS20997G25
ACTIAS20997G100EA 0 CHF
ACTIAS20997G100 ACTIAS20997G25
(R)-(-)-Methylmandelat
(R)-(-)-Methylmandelat
Formel: C₉H₁₀O₃
Siedepunkt: 267 °C (1013 hPa)
Melting Pt: 51…53 °C
MDL Number: MFCD00064247
CAS-Nummer: 20698-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR