D(+)-Homoserin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: H-D-Hser-OH, H-D-HomoSer-OH, H-D-Homoser-OH, (R)-(+)-Homoserine, (R)-2-Amino-4-hydroxybutyric acid, D-Homoserine

AS31501G1 AS31501G5
ACTIAS31501G1EA 0 CHF
ACTIAS31501G1 ACTIAS31501G5
D(+)-Homoserin
D(+)-Homoserin
Formel: C₄H₉NO₃
MW: 119,12 g/mol
MDL Number: MFCD00077786
CAS-Nummer: 6027-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR