(±)-2-Phenylpiperazin
Lieferant: ACTIVATE SCIENTIFIC
AS30075G1
AS30075G5
ACTIAS30075G1EA
0
CHF
ACTIAS30075G1
ACTIAS30075G5
(±)-2-Phenylpiperazin
(±)-2-Phenylpiperazin
Formel:
C₁₀H₁₄N₂ MW: 162,23 g/mol |
MDL Number:
MFCD01871362 CAS-Nummer: 5271-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...