4-Hydroxy-3-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Nitro-4-pyridon, 3-Nitro-4-pyridone, 3-Nitropyridin-4-ol, 3-Nitro-4-pyridinol

AS21320G100 AS21320G25
ACTIAS21320G100EA 0 CHF
ACTIAS21320G100 ACTIAS21320G25
4-Hydroxy-3-nitropyridin
4-Hydroxy-3-nitropyridine
Formel: C₅H₄N₂O₃
MW: 140,10 g/mol
MDL Number: MFCD00128874
CAS-Nummer: 5435-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR