5-Chlor-2-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33125G1 AS33125G5
ACTIAS33125G1EA 0 CHF
ACTIAS33125G1 ACTIAS33125G5
5-Chlor-2-nitropyridin
5-Chlor-2-nitropyridin
Formel: C₅H₃ClN₂O₂
MW: 158,54 g/mol
MDL Number: MFCD03092916
CAS-Nummer: 52092-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR