3-Chlor-2-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS31387G1 AS31387G5
ACTIAS31387G1EA 0 CHF
ACTIAS31387G1 ACTIAS31387G5
3-Chlor-2-nitropyridin
3-Chloro-2-nitropyridine
Formel: C₅H₃ClN₂O₂
MDL Number: MFCD00490110
CAS-Nummer: 54231-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR