2-Methoxy-3-methylpyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS24533G100 AS24533G25
ACTIAS24533G100EA 0 CHF
ACTIAS24533G100 ACTIAS24533G25
2-Methoxy-3-methylpyrazin
2-Methoxy-3-methylpyrazine
Formel: C₆H₈N₂O
MW: 124.14 g/mol
Siedepunkt: 87 °C (60 mmHg)
Dichte: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00006127
CAS-Nummer: 2847-30-5
EINECS: 220-651-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR