2-Iodpyrazin
Lieferant: ACTIVATE SCIENTIFIC
AS1946G25
AS1946G5
ACTIAS1946G25EA
0
CHF
ACTIAS1946G25
ACTIAS1946G5
2-Iodpyrazin
2-Iodopyrazine
Formel:
C₄H₃IN₂ |
MDL Number:
MFCD01319019 CAS-Nummer: 32111-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...