4-Brom-1-naphthylamin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
1-Amino-4-bromonaphthalene
AS20410G100
AS20410G25
ACTIAS20410G100EA
0
CHF
ACTIAS20410G100
ACTIAS20410G25
4-Brom-1-naphthylamin
4-Brom-1-naphthylamin
Formel:
C₁₀H₈BrN MW: 222,08 g/mol |
MDL Number:
MFCD00004023 CAS-Nummer: 2298-07-9 EINECS: 218-944-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...