1-(3-Nitrophenyl)piperazin

Lieferant: ACTIVATE SCIENTIFIC

AS35124G1 AS35124G5
ACTIAS35124G1EA 0 CHF
ACTIAS35124G1 ACTIAS35124G5
1-(3-Nitrophenyl)piperazin
1-(3-Nitrophenyl)piperazin
Formel: C₁₀H₁₃N₃O₂
CAS-Nummer: 54054-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR