4'-Fluor-2'-hydroxyacetophenon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Fluoro-2-hydroxyacetophenone

AS35910G1 AS35910G5
ACTIAS35910G1EA 0 CHF
ACTIAS35910G1 ACTIAS35910G5
4'-Fluor-2'-hydroxyacetophenon
4'-Fluor-2'-hydroxyacetophenon
Formel: C₈H₇FO₂
MDL Number: MFCD00143203
CAS-Nummer: 1481-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR