5-Methoxy-2-nitrophenol

Lieferant: ACTIVATE SCIENTIFIC

AS35369G1 AS35369G10 AS35369G5
ACTIAS35369G1EA 0 CHF
ACTIAS35369G1 ACTIAS35369G10 ACTIAS35369G5
5-Methoxy-2-nitrophenol
5-Methoxy-2-nitrophenol
Formel: C₇H₇NO₄
MW: 169,14 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00100932
CAS-Nummer: 704-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR