4-Fluor-2-iodanilin
Lieferant: ACTIVATE SCIENTIFIC
AS22394G100
AS22394G25
ACTIAS22394G100EA
188
CHF
ACTIAS22394G100
ACTIAS22394G25
4-Fluor-2-iodanilin
4-Fluor-2-iodanilin
Formel:
C₆H₅FIN MW: 237,02 g/mol |
MDL Number:
MFCD00042584 CAS-Nummer: 61272-76-2 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...