5-Fluor-2-iodanilin
Lieferant: ACTIVATE SCIENTIFIC
AS35206G1
AS35206G10
AS35206G5
ACTIAS35206G1EA
0
CHF
ACTIAS35206G1
ACTIAS35206G10
ACTIAS35206G5
5-Fluor-2-iodanilin
5-Fluor-2-iodanilin
Formel:
C₆H₅FIN MW: 237,02 g/mol Siedepunkt: 259 °C (1013 hPa) Melting Pt: 40…42 °C |
MDL Number:
MFCD02093953 CAS-Nummer: 255724-71-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...