2-Chlor-4-methoxyphenol
Lieferant: ACTIVATE SCIENTIFIC
AS33725G100
AS33725G5
ACTIAS33725G100EA
0
CHF
ACTIAS33725G100
ACTIAS33725G5
2-Chlor-4-methoxyphenol
2-Chlor-4-methoxyphenol
Formel:
C₇H₇ClO₂ Siedepunkt: 242 °C (1013 hPa) Melting Pt: 46…47 °C |
MDL Number:
MFCD00070773 CAS-Nummer: 18113-03-6 EINECS: 242-007-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...