4-Chlor-2-(trifluormethoxy)anilin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Chloro-2-(trifluoromethoxy)phenylamine

AS35100G1 AS35100G5
ACTIAS35100G1EA 0 CHF
ACTIAS35100G1 ACTIAS35100G5
4-Chlor-2-(trifluormethoxy)anilin
4-Chlor-2-(trifluormethoxy)anilin
Formel: C₇H₅ClF₃NO
MW: 211.57 g/mol
MDL Number: MFCD00153102
CAS-Nummer: 175205-77-3
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR