3-Chlor-4-(trifluormethoxy)anilin

Lieferant: ACTIVATE SCIENTIFIC

AS34857G10 AS34857G100 AS34857G5
ACTIAS34857G10EA 0 CHF
ACTIAS34857G10 ACTIAS34857G100 ACTIAS34857G5
3-Chlor-4-(trifluormethoxy)anilin
3-Chlor-4-(trifluormethoxy)anilin
Formel: C₇H₅ClF₃NO
MW: 211.57 g/mol
Siedepunkt: 241
Melting Pt: 29…30 °C
Dichte: 1.454 g/cm³ (20 °C)
MDL Number: MFCD00190126
CAS-Nummer: 64628-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR