4-Brom-3-methoxyphenol

Lieferant: ACTIVATE SCIENTIFIC

AS35244G1 AS35244G5
ACTIAS35244G1EA 0 CHF
ACTIAS35244G1 ACTIAS35244G5
4-Brom-3-methoxyphenol
4-Brom-3-methoxyphenol
Formel: C₇H₇BrO₂
MW: 203.04 g/mol
MDL Number: MFCD00029748
CAS-Nummer: 102127-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR