2-Brom-6-nitrophenol

Lieferant: ACTIVATE SCIENTIFIC

AS21247G100 AS21247G25 AS21247G5
ACTIAS21247G100EA 0 CHF
ACTIAS21247G100 ACTIAS21247G25 ACTIAS21247G5
2-Brom-6-nitrophenol
2-Brom-6-nitrophenol
Formel: C₆H₄BrNO₃
MW: 218.01 g/mol
MDL Number: MFCD02683216
CAS-Nummer: 13073-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR