4-Brom-3-nitroanilin
Lieferant: ACTIVATE SCIENTIFIC
AS33357G1
AS33357G5
ACTIAS33357G1EA
0
CHF
ACTIAS33357G1
ACTIAS33357G5
4-Brom-3-nitroanilin
4-Brom-3-nitroanilin
Formel:
C₆H₅BrN₂O₂ MW: 217,02 g/mol |
MDL Number:
MFCD01320678 CAS-Nummer: 53324-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...