3-Brom-4-(trifluormethoxy)anilin

Lieferant: ACTIVATE SCIENTIFIC

AS33731G1 AS33731G10 AS33731G5
ACTIAS33731G1EA 0 CHF
ACTIAS33731G1 ACTIAS33731G10 ACTIAS33731G5
3-Brom-4-(trifluormethoxy)anilin
3-Brom-4-(trifluormethoxy)anilin
Formel: C₇H₅BrF₃NO
MW: 256.02 g/mol
MDL Number: MFCD00190127
CAS-Nummer: 191602-54-7
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR