2-Brom-5-methylthiophen

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Methyl-2-bromothiophene, 2-Bromo-5-methylthiophenol

AS20197G100 AS20197G25
ACTIAS20197G100EA 126 CHF
ACTIAS20197G100 ACTIAS20197G25
2-Brom-5-methylthiophen
2-Bromo-5-methylthiophene
Formel: C₅H₅BrS
MW: 177,06 g/mol
Siedepunkt: 178 °C (1013 hPa)
Melting Pt: 58 °C
Dichte: 1,5529 g/cm³ (20 °C)
MDL Number: MFCD00130103
CAS-Nummer: 765-58-2
EINECS: 212-151-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR