3-Acetylpyrrol
Lieferant: ACTIVATE SCIENTIFIC
AS36533G1
AS36533G5
ACTIAS36533G1EA
0
CHF
ACTIAS36533G1
ACTIAS36533G5
3-Acetylpyrrol
3-Acetylpyrrol
Formel:
C₆H₇NO |
MDL Number:
MFCD00067759 CAS-Nummer: 1072-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...