2-Fluornicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoropyridine-3-carboxylic acid, 2-Fluoro-3-pyridinecarboxylic acid

AS21307G25 AS21307G5
ACTIAS21307G25EA 81 CHF
ACTIAS21307G25 ACTIAS21307G5
2-Fluornicotinsäure
2-Fluornicotinsäure
Formel: C₆H₄FNO₂
MW: 141,1 g/mol
MDL Number: MFCD00040744
CAS-Nummer: 393-55-5
EINECS: 206-888-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR