Platin(II)chlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Platinum dichloride

AS23498G1 AS23498G5
ACTIAS23498G1EA 0 CHF
ACTIAS23498G1 ACTIAS23498G5
Platin(II)chlorid
Platin(II)chlorid
Formel: Cl₂Pt
MW: 265,99 g/mol
Melting Pt: 581 °C
Dichte: 6.05 g/cm³ (20 °C)
MDL Number: MFCD00011181
CAS-Nummer: 10025-65-7
EINECS: 233-034-1
UN: 3260
ADR: 8,III
Merck Index: 12,07687

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR