4-Fluor-3-nitrotoluol

Lieferant: ACTIVATE SCIENTIFIC

AS33990G10 AS33990G100
ACTIAS33990G10EA 0 CHF
ACTIAS33990G10 ACTIAS33990G100
4-Fluor-3-nitrotoluol
4-Fluor-3-nitrotoluol
Formel: C₇H₆FNO₂
MDL Number: MFCD00007060
CAS-Nummer: 446-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR