5-Amino-α,α,α-trifluor-m-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC

AS35667G1 AS35667G5
ACTIAS35667G1EA 0 CHF
ACTIAS35667G1 ACTIAS35667G5
5-Amino-α,α,α-trifluor-m-toluylsäure
5-Amino-α,α,α-trifluor-m-toluylsäure
Formel: C₈H₆F₃NO₂
MW: 205,14 g/mol
MDL Number: MFCD00236641
CAS-Nummer: 328-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR